Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173374.1 | NP_001166845.1 | 4, 5 | 631 | ||
GenBank mRNA | BC158882.1 | - | 4, 5 | 566 |