Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109035.1 | NP_001102505.1 | 4 | 609 | ||
XM_006246786.2 | XP_006246848.1 | 5 | 1198 | |||
XM_006246789.2 | XP_006246851.1 | 5 | 1199 | |||
XM_008767857.1 | XP_008766079.1 | 5 | 1199 | |||
XM_008767858.1 | XP_008766080.1 | 5 | 1199 |