Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191555.1 | NP_001178484.1 | 6 | 3520 | ||
NM_001197241.1 | NP_001184170.1 | 5 | 1897 | |||
XM_008771331.1 | XP_008769553.1 | 6 | 3777 | |||
GenBank mRNA | BC107469.1 | - | 5 | 1897 | 7458514 | |
FQ221846.1 | - | 5 | 1906 |