Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253849.2 | XP_006253911.1 | 3, 4 | 525 | ||
GenBank mRNA | FQ210031.1 | - | 3, 4 | 397 | ||
FQ227425.1 | - | 3, 4 | 393 | |||
FQ229485.1 | - | 3, 4 | 407 |