Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033923.1 | NP_001029095.1 | 4 | 1390 | ||
XM_006236441.2 | XP_006236503.1 | 5 | 1367 | |||
GenBank mRNA | AJ633685.1 | - | 4 | 1231 | ||
BC099228.1 | - | 4 | 1390 | 7389353 | ||
DQ125335.1 | - | 4 | 1420 | |||
DQ125336.1 | - | 4 | 1420 |