Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126086.2 | NP_001119558.1 | 3 | 210 | ||
NM_001145449.1 | NP_001138921.1 | 3 | 185 | |||
XM_006239651.2 | XP_006239713.2 | 3 | 1265 | |||
GenBank mRNA | BC166893.1 | - | 3 | 189 | 6919817 | |
CB765303.1 | - | 3 | 185 | |||
FM039224.1 | - | 3 | 173 |