Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4dl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765998.1 | XP_008764220.1 | 2 | 2186 | ||
XM_008765999.1 | XP_008764221.1 | 2 | 2189 |