Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109328.2 | NP_001102798.1 | 2 | 127 | ||
XM_008769490.1 | XP_008767712.1 | 2 | 196 | |||
XM_008769491.1 | XP_008767713.1 | 2 | 542 | |||
XM_008769492.1 | XP_008767714.1 | 2 | 543 | |||
XM_008769493.1 | XP_008767715.1 | 2 | 547 | |||
GenBank mRNA | FM076322.1 | - | 2 | 127 |