Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109333.1 | NP_001102803.1 | 2 | 1167 | ||
GenBank mRNA | FQ217859.1 | - | 2 | 1227 |