Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024370.2 | NP_001019541.1 | 2 | 301 | ||
GenBank mRNA | BC086361.1 | - | 2 | 301 | 7304702 |