Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020103.1 | NP_064488.1 | 3 | 245 | ||
XM_006257716.2 | XP_006257778.1 | 4 | 350 | |||
GenBank mRNA | FQ230231.1 | - | 4 | 303 | ||
FQ230335.1 | - | 4 | 304 | |||
FQ232627.1 | - | 4 | 304 | |||
FQ234379.1 | - | 4 | 303 | |||
FQ235068.1 | - | 4 | 304 | |||
M30691.1 | - | 3 | 243 |
Gene Symbol: LOC100911104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751343.2 | XP_003751391.1 | 4 | 350 |