Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031615.1 | NP_113803.1 | 9 | 2561 | ||
XM_006248435.2 | XP_006248497.1 | 6 | 2209 | |||
GenBank mRNA | U30381.1 | - | 9 | 2561 |