Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019383.2 | NP_062256.1 | 5 | 382 | ||
XM_008768390.1 | XP_008766612.1 | 4 | 347 | |||
GenBank mRNA | BC059139.1 | - | 5 | 386 | 6921557 | |
BC078846.1 | - | 4 | 334 | 7133871 | ||
D10021.1 | - | 4 | 354 | |||
D13120.1 | - | 5 | 375 | |||
FQ211219.1 | - | 5 | 393 | |||
FQ216885.1 | - | 5 | 382 | |||
FQ217843.1 | - | 5 | 393 | |||
FQ222479.1 | - | 5 | 375 |