Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022962.1 | NP_075251.1 | 13 | 2585 | ||
GenBank mRNA | AF081144.1 | - | 12 | 2570 | ||
AF081145.1 | - | 12 | 2570 | |||
AF081146.1 | - | 13 | 2585 | |||
AF081147.1 | - | 13 | 2585 | |||
AF111099.1 | - | 14 | 2911 | |||
U72487.1 | - | 12 | 2387 | |||
U78105.1 | - | 12 | 2712 |
Set Membership: |
G-protein coupled receptor |