Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242722.2 | XP_006242784.1 | 21 | 2185 | ||
XM_235972.8 | XP_235972.6 | 20 | 2134 |