Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766891.1 | XP_008765113.1 | 3 | 508 | ||
XM_008766892.1 | XP_008765114.1 | 3 | 446 | |||
XM_008766893.1 | XP_008765115.1 | 2 | 107 | |||
XM_008766894.1 | XP_008765116.1 | 2 | 260 |