Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100806.1 | NP_001094276.1 | 1 | 827 | ||
GenBank mRNA | BC168678.1 | - | 1 | 827 | 5598674 |