Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gypa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001069014.4 | XP_001069014.1 | 7 | 481 | ||
XM_006255410.2 | XP_006255472.1 | 6 | 388 |