Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023982.1 | NP_076472.1 | 13 | 942 | ||
XM_006232759.1 | XP_006232821.1 | 14 | 2204 | |||
XM_006232766.1 | XP_006232828.1 | 12 | 2156 | |||
XM_008761214.1 | XP_008759436.1 | 12 | 2143 | |||
XM_008761215.1 | XP_008759437.1 | 12 | 2143 | |||
XM_008761216.1 | XP_008759438.1 | 12 | 2140 | |||
XM_008761217.1 | XP_008759439.1 | 12 | 2140 | |||
XM_008761218.1 | XP_008759440.1 | 13 | 3117 | |||
XM_008761219.1 | XP_008759441.1 | 12 | 2143 | |||
XM_008761220.1 | XP_008759442.1 | 12 | 2140 | |||
XM_008761221.1 | XP_008759443.1 | 12 | 3102 | |||
XM_008761222.1 | XP_008759444.1 | 11 | 2023 | |||
XM_008761223.1 | XP_008759445.1 | 12 | 2143 | |||
XM_008761224.1 | XP_008759446.1 | 11 | 2020 | |||
XM_008761225.1 | XP_008759447.1 | 11 | 2005 | |||
XM_008761226.1 | XP_008759448.1 | 12 | 2997 | |||
GenBank mRNA | AF225961.1 | - | 13 | 942 |