Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031004.2 | NP_112266.1 | 6 | 612 | ||
GenBank mRNA | BC158550.1 | - | 6 | 606 | 9033322 | |
CB608976.1 | - | 2 | 207 | |||
DN934985.1 | - | 6 | 641 | |||
X06801.1 | - | 6 | 597 |