Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146156.1 | NP_001139628.1 | 3 | 1284 | ||
NM_002093.3 | NP_002084.2 | 3 | 1284 | |||
XM_006713610.2 | XP_006713673.1 | 3 | 770 | |||
XM_006713611.2 | XP_006713674.1 | 3 | 770 | |||
GenBank mRNA | AB451356.1 | - | 3 | 301 | ||
AK290897.1 | - | 3 | 657 | |||
BC000251.1 | - | 3 | 533 | 3357620 | ||
BC012760.2 | - | 3 | 1283 | 3637163 | ||
CR536510.1 | - | 3 | 301 | |||
EU302497.1 | - | 3 | 301 | |||
L33801.1 | - | 3 | 340 |