Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017572.3 | NP_060042.2 | 14 | 1473 | ||
NM_199054.2 | NP_951009.1 | 14 | 3500 | |||
GenBank mRNA | AB209074.1 | - | 4 | 3810 | ||
AF125532.1 | - | 13 | 1250 | |||
AF237776.1 | - | 13 | 1264 | |||
AK001730.1 | - | 4 | 524 | |||
AK023442.1 | - | 1 | 967 | |||
AK024347.1 | - | 1 | 1700 | |||
AK092536.1 | - | 4 | 2403 | |||
AK097012.1 | - | 1 | 2311 | |||
AL137565.1 | - | 10 | 935 | |||
AL137615.1 | - | 1 | 1184 | |||
BC018345.1 | - | 6 | 642 | |||
BC021235.2 | - | 2 | 2158 | |||
BC073140.1 | - | 14 | 1453 | 6527053 | ||
BM977403.1 | - | 1 | 300 |