Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012324.4 | NP_036456.1 | 10, 11 | 2438 | ||
XM_011530679.2 | XP_011528981.1 | 10, 11 | 2330 | |||
XM_011530680.2 | XP_011528982.1 | 10, 11 | 2216 | |||
XM_011530681.2 | XP_011528983.1 | 10, 11 | 2195 | |||
GenBank mRNA | AF136382.1 | - | 10, 11 | 2421 | ||
AF218778.1 | - | 8, 9 | 2223 | |||
AK293580.1 | - | 8, 9 | 809 | |||
AK294265.1 | - | 11, 12 | 1497 | |||
AL021708.1 | - | 6, 7 | 1441 | |||
BC009940.2 | - | 9, 10 | 1188 | 4100637 | ||
BC047527.1 | - | 10, 11 | 2351 | 5277237 | ||
U79261.1 | - | 5, 6 | 512 |