Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243835.1 | NP_001230764.1 | 4 | 566 | ||
NM_003151.3 | NP_003142.1 | 4 | 617 | |||
XM_006712719.3 | XP_006712782.1 | 4 | 612 | |||
XM_011511705.2 | XP_011510007.1 | 7 | 763 | |||
XM_017004784.1 | XP_016860273.1 | 4 | 371 | |||
GenBank mRNA | AK299976.1 | - | 4 | 338 | ||
AK300868.1 | - | 4 | 370 | |||
BC031212.1 | - | 4 | 538 | 4830583 | ||
BF966132.1 | - | 4 | 565 | |||
L78440.1 | - | 4 | 383 |