Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX2OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002791.2 | 4 | 3461 | |||
GenBank mRNA | AL137578.1 | - | 1 | 409 | ||
AY117034.1 | - | 4 | 3461 | |||
AY117413.1 | - | LIT2056 EMX2OS |
4 | 3287 | ||
BC040186.2 | - | 3 | 3126 |