Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-CE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002155.1 | 1 | 653 |
Gene Symbol: RT1-CE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC083874.1 | - | 2 | 456 |