Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mirg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_028265.1 | 3, 4 | 299 | |||
GenBank mRNA | AI605771.1 | - | 3, 4 | 299 | ||
AJ517767.1 | - | LIT1749 Mirg |
3, 4 | 299 | ||
EU434920.1 | - | 2184 | ||||
EU616812.1 | - | 2883 | ||||
EU616813.1 | - | 2796 |