Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mirg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_028265.1 | 6 | 735 | |||
GenBank mRNA | AJ517767.1 | - | LIT1749 Mirg |
6 | 735 | |
AK013406.1 | - | 1 | 32 | |||
EU434920.1 | - | 2620 | ||||
EU616812.1 | - | 3257 | ||||
EU616813.1 | - | 3232 |