Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027876.1 | 5, 6 | 712 | |||
GenBank mRNA | AK088008.1 | - | 5, 6 | 577 |