Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox2ot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015580.2 | 5 | 930 | |||
GenBank mRNA | AK031826.1 | - | 2 | 492 | ||
AK031919.1 | - | 674 | ||||
AK045614.1 | - | 3 | 668 | |||
BC057611.1 | - | 5 | 930 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CX563303.1 | - | 3 | 668 | Chr. 3 - 34638364 - 34676045 [+] on Build GRCm38 |