Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vax2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002873.2 | 2 | 1835 | |||
GenBank mRNA | AY589787.1 | - | LIT3407 Vax2OS |
2 | 1839 |