Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002864.1 | 1 | 3686 | |||
GenBank mRNA | AY151252.1 | - | LIT1829 Peg13 |
1 | 3686 | |
AY151253.1 | - | LIT1828 Peg13 |
1 | 3360 |