Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx1as | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002854.2 | 2 | 1192 | |||
GenBank mRNA | AK132348.1 | - | 2 | 1193 |
Gene Symbol: Dlx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138311.1 | - | 1195 |