Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930526I15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015516.2 | 1143 | ||||
NR_027950.2 | 2 | 781 | ||||
NR_102721.1 | 1024 | |||||
GenBank mRNA | AK015904.1 | - | 1024 | |||
AK078907.1 | - | 1 | 855 |