Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nespas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002846.2 | 4, 5 | 980 | |||
GenBank mRNA | AF319945.1 | - | LIT1669 Nespas |
4, 5 | 951 | |
AK153813.1 | - | 4, 5 | 981 |