Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snora65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002898.2 | 1 | 80 | |||
GenBank mRNA | AF357418.1 | - | 1 | 80 |
Gene Symbol: Rpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148566.1 | - | 898 | |||
BC075731.1 | - | 3 | 885 |