Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vaultrc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027885.1 | 1 | 76 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AF210457.1 | - | 1 | 76 | Chr. - - [-] on Build |