Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r-ps60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028441.1 | 1 | 38 | |||
GenBank mRNA | AF022253.1 | - | 1 | 38 |
Gene Symbol: Vmn2r-ps57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF022250.1 | - | 1 | 186 |