Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930433N12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027988.1 | 3 | 475 | |||
GenBank mRNA | AK015306.1 | - | 353 | |||
AK076785.1 | - | 3 | 442 | |||
BB614377.1 | - | 3 | 477 | |||
BC061047.1 | - | 2 | 370 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_053265.1 | - | 353 | Chr. - - [-] on Build |