Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027994.1 | 3 | 1290 | |||
GenBank mRNA | BC025319.1 | - | 3 | 1295 | ||
BC033111.1 | - | 3 | 1295 | |||
BC080649.1 | - | 3 | 1295 |