Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSORS1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026816.1 | 3 | 356 | |||
GenBank mRNA | AB932952.1 | - | 2 | 323 | ||
AB932954.1 | - | 3 | 354 | |||
AB933312.1 | - | 4 | 621 | |||
AB933313.1 | - | 4 | 387 | |||
AB933320.1 | - | 6 | 839 | |||
AB933321.1 | - | 4 | 535 | |||
AY484516.1 | - | 3 | 356 | |||
LC027944.1 | - | 3 | 590 | |||
LC027945.1 | - | 4 | 648 | |||
LC027946.1 | - | 3 | 615 | |||
LC050981.1 | - | 5 | 832 | |||
LC050982.1 | - | 5 | 913 | |||
LC050983.1 | - | 5 | 679 | |||
LC050984.1 | - | 3 | 387 | |||
LC050985.1 | - | 4 | 508 | |||
LC050986.1 | - | 6 | 865 | |||
LC050987.1 | - | 7 | 1132 | |||
LC050988.1 | - | 2 | 354 | |||
LC050989.1 | - | 4 | 646 | |||
LC050990.1 | - | 6 | 718 | |||
LC050991.1 | - | 4 | 882 |