Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSORS1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026816.1 | 3 | 387 | |||
GenBank mRNA | AB932952.1 | - | 2 | 354 | ||
AB932954.1 | - | 3 | 385 | |||
AB933312.1 | - | 4 | 652 | |||
AB933313.1 | - | 4 | 418 | |||
AB933320.1 | - | 6 | 870 | |||
AB933321.1 | - | 4 | 566 | |||
AY484516.1 | - | 3 | 387 | |||
LC027944.1 | - | 3 | 621 | |||
LC027945.1 | - | 4 | 679 | |||
LC027946.1 | - | 3 | 646 | |||
LC050981.1 | - | 5 | 863 | |||
LC050982.1 | - | 5 | 944 | |||
LC050983.1 | - | 5 | 710 | |||
LC050984.1 | - | 3 | 418 | |||
LC050985.1 | - | 4 | 539 | |||
LC050986.1 | - | 6 | 896 | |||
LC050987.1 | - | 7 | 1163 | |||
LC050988.1 | - | 2 | 385 | |||
LC050989.1 | - | 4 | 677 | |||
LC050990.1 | - | 6 | 749 | |||
LC050991.1 | - | 4 | 913 |