Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSORS1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026816.1 | 2 | 277 | |||
GenBank mRNA | AB933313.1 | - | 3 | 308 | ||
AB933320.1 | - | 5 | 760 | |||
AB933321.1 | - | 3 | 456 | |||
AY484516.1 | - | 2 | 277 | |||
LC027945.1 | - | 3 | 569 | |||
LC050983.1 | - | 4 | 600 | |||
LC050984.1 | - | 2 | 308 | |||
LC050985.1 | - | 3 | 429 | |||
LC050986.1 | - | 5 | 786 | |||
LC050987.1 | - | 6 | 1053 | |||
LC050990.1 | - | 5 | 639 |