Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KLKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002948.1 | 5 | 945 | |||
GenBank mRNA | AF355199.1 | - | 2 | 507 | ||
AF464754.1 | - | 2 | 683 | |||
AY533562.1 | - | 5 | 945 | |||
DQ086829.1 | - | 2 | 1352 | |||
DQ211698.1 | - | 5 | 2668 |