Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNRHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002328.4 | 1 | 261 | |||
NR_104034.1 | 1 | 84 | ||||
GenBank mRNA | AB239331.1 | - | 1 | 84 | ||
AK310736.1 | - | 1 | 261 | |||
AY077708.1 | - | 1 | 261 | |||
AY233132.1 | - | 1 | 255 | |||
AY233133.1 | - | 1 | 30 | |||
DQ266410.1 | - | 1 | 84 |