Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R3CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002784.2 | 1 | 1367 | |||
GenBank mRNA | AK057429.1 | - | 1 | 442 | ||
AK131475.1 | - | 1 | 1367 | |||
DB022709.1 | - | 1 | 442 |