Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFG3L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003226.1 | 9 | 1027 | |||
NR_003227.1 | 8 | 854 | ||||
NR_003228.1 | 9 | 993 | ||||
GenBank mRNA | AF329691.1 | - | 8 | 778 | ||
AF329692.1 | - | 3 | 329 | |||
AK056488.1 | - | 9 | 998 | |||
AK091293.1 | - | 9 | 990 | |||
AK097044.1 | - | 8 | 841 | |||
AK126911.1 | - | 7 | 818 | |||
AK128352.1 | - | 2 | 693 | |||
AL832984.1 | - | 3 | 251 | |||
BC021245.2 | - | 7 | 600 |