Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026597.1 | 8, 9 | 2288 | |||
GenBank mRNA | AK024261.1 | - | 7, 8 | 1456 | ||
AK090954.1 | - | 8, 9 | 2291 |