Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAX SIRNA Assays ›
Gene Symbol:
BAX
Gene Name:
BCL2 associated X, apoptosis regulator
Gene Aliases:
BCL2L4
Chromosome Location:
Chr.19: 48954825 - 48961798 on Build GRCh38
UniGene ID:
Hs.624291
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n259082
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027882.1
5
450
GenBank mRNA
AF007826.1
-
5
381
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
neuron migration
T cell homeostatic proliferation
B cell homeostasis
B cell apoptotic process
kidney development
release of cytochrome c from mitochondria
protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
blood vessel remodeling
myeloid cell homeostasis
B cell negative selection
B cell homeostatic proliferation
positive regulation of B cell apoptotic process
glycosphingolipid metabolic process
regulation of nitrogen utilization
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
activation of signaling protein activity involved in unfolded protein response
inner mitochondrial membrane organization
outer mitochondrial membrane organization
germ cell development
aging
mitochondrial fusion
extrinsic apoptotic signaling pathway via death domain receptors
intrinsic apoptotic signaling pathway in response to DNA damage
activation of cysteine-type endopeptidase activity involved in apoptotic process by cytochrome c
apoptotic mitochondrial changes
fertilization
response to toxic substance
response to salt stress
establishment or maintenance of transmembrane electrochemical gradient
response to gamma radiation
viral process
hypothalamus development
cerebral cortex development
negative regulation of protein binding
positive regulation of protein oligomerization
endoplasmic reticulum calcium ion homeostasis
negative regulation of endoplasmic reticulum calcium ion concentration
release of matrix enzymes from mitochondria
negative regulation of peptidyl-serine phosphorylation
regulation of mammary gland epithelial cell proliferation
glial cell apoptotic process
cellular response to UV
ectopic germ cell programmed cell death
response to cocaine
odontogenesis of dentin-containing tooth
response to drug
regulation of apoptotic process
positive regulation of apoptotic process
regulation of protein homodimerization activity
regulation of protein heterodimerization activity
negative regulation of neuron apoptotic process
positive regulation of neuron apoptotic process
mitochondrial fragmentation involved in apoptotic process
development of secondary sexual characteristics
cellular respiration
retinal cell programmed cell death
response to copper ion
positive regulation of developmental pigmentation
negative regulation of fibroblast proliferation
spermatid differentiation
post-embryonic camera-type eye morphogenesis
response to axon injury
homeostasis of number of cells within a tissue
protein oligomerization
protein homooligomerization
positive regulation of release of sequestered calcium ion into cytosol
neuron apoptotic process
response to corticosterone
regulation of mitochondrial membrane potential
Sertoli cell proliferation
retina development in camera-type eye
positive regulation of apoptotic process involved in mammary gland involution
vagina development
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
thymocyte apoptotic process
mitochondrion morphogenesis
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of release of cytochrome c from mitochondria
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
intrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
positive regulation of endoplasmic reticulum unfolded protein response
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
apoptotic process involved in patterning of blood vessels
apoptotic process involved in embryonic digit morphogenesis
regulation of mitochondrial membrane permeability involved in programmed necrotic cell death
positive regulation of apoptotic DNA fragmentation
positive regulation of IRE1-mediated unfolded protein response
B cell receptor apoptotic signaling pathway
negative regulation of apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
positive regulation of intrinsic apoptotic signaling pathway
protein binding
lipid binding
channel activity
heat shock protein binding
protein complex binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
chaperone binding
BH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0