Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR9-3HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015411.1 | 7 | 2370 | |||
NR_133001.1 | 7 | 2950 | ||||
NR_133002.1 | 6 | 2251 | ||||
NR_133003.1 | 7 | 2285 | ||||
GenBank mRNA | AK096359.1 | - | 1 | 486 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA130217.1 | - | 1 | 36 | Chr. 15 - 89396624 - 89397184 [+] on Build GRCh38 |